Assay Details
Gene Symbol: Gabrd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017289.1 | NP_058985.1 | 4 - 5 | 518 | 71 | |
GenBank mRNA | BC087714.1 | - | 4 - 5 | 483 | 7317154 | 71 |
M35162.1 | - | 4 - 5 | 518 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
