Assay Details
Gene Symbol: Ahsa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115034.1 | NP_001108506.1 | 6 - 7 | 829 | 64 | |
GenBank mRNA | BC158698.1 | - | 6 - 7 | 786 | 9027714 | 64 |
FQ223544.1 | - | 6 - 7 | 813 | 64 | ||
FQ233084.1 | - | 6 - 7 | 791 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
