Assay Details
Gene Symbol: Gstz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109445.1 | NP_001102915.1 | 4 - 5 | 343 | 77 | |
XM_006240399.2 | XP_006240461.1 | 4 - 5 | 317 | 77 | ||
GenBank mRNA | BC158833.1 | - | 4 - 5 | 285 | 7381517 | 77 |
FJ179410.1 | - | 3 - 4 | 220 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
