Assay Details
Gene Symbol: Gstz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109445.1 | NP_001102915.1 | 1 - 2 | 138 | 63 | |
GenBank mRNA | BC158833.1 | - | 1 - 2 | 80 | 63 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC158833.1, NM_001109445.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
