Assay Details
Gene Symbol: Gstz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006240399.2 | XP_006240461.1 | 1 - 2 | 110 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006240399.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
