Assay Details
Gene Symbol: Coq6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011983.1 | NP_001011983.1 | 10 - 11 | 1257 | 70 | |
GenBank mRNA | BC079342.1 | - | 10 - 11 | 1257 | 70 | |
FQ228932.1 | - | 9 - 10 | 1614 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
