Assay Details
Gene Symbol: Smoc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002835.1 | NP_001002835.1 | 7 - 8 | 659 | 111 | |
XM_006240284.2 | XP_006240346.1 | 7 - 8 | 965 | 111 | ||
XM_006240285.2 | XP_006240347.1 | 7 - 8 | 965 | 111 | ||
XM_008764785.1 | XP_008763007.1 | 8 - 9 | 1065 | 111 | ||
XM_008764786.1 | XP_008763008.1 | 8 - 9 | 1032 | 111 | ||
GenBank mRNA | FQ222878.1 | - | 7 - 8 | 916 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
