Assay Details
Gene Symbol: Tc2n | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025152.1 | NP_001020323.1 | 4 - 5 | 356 | 123 | |
XM_008764831.1 | XP_008763053.1 | 3 - 4 | 2019 | 123 | ||
XM_008764832.1 | XP_008763054.1 | 4 - 5 | 1668 | 123 | ||
XM_008764833.1 | XP_008763055.1 | 4 - 5 | 1915 | 123 | ||
XM_008764834.1 | XP_008763056.1 | 3 - 4 | 889 | 123 | ||
XM_008764835.1 | XP_008763057.1 | 4 - 5 | 920 | 123 | ||
XM_008764836.1 | XP_008763058.1 | 4 - 5 | 538 | 123 | ||
XM_008764837.1 | XP_008763059.1 | 4 - 5 | 883 | 123 | ||
XM_008764838.1 | XP_008763060.1 | 3 - 4 | 852 | 123 | ||
XM_008764839.1 | XP_008763061.1 | 3 - 4 | 418 | 123 | ||
XM_008764840.1 | XP_008763062.1 | 2 - 3 | 285 | 123 | ||
GenBank mRNA | BC088432.1 | - | 4 - 5 | 356 | 123 | |
FQ211170.1 | - | 4 - 5 | 474 | 123 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC088432.1, NM_001025152.1, XM_008764831.1, XM_008764832.1, XM_008764833.1, XM_008764834.1, XM_008764835.1, XM_008764836.1, XM_008764837.1, XM_008764838.1, XM_008764839.1, XM_008764840.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
