Assay Details
Gene Symbol: Aldh6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031057.2 | NP_112319.2 | 2 - 3 | 160 | 107 | |
XM_006240354.2 | XP_006240416.1 | 2 - 3 | 525 | 107 | ||
XM_008764863.1 | XP_008763085.1 | 2 - 3 | 162 | 107 | ||
GenBank mRNA | DN948946.1 | - | 2 - 3 | 157 | 107 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008764863.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
