Assay Details
Gene Symbol: Noxred1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750205.3 | XP_003750253.1 | 4 - 5 | 835 | 64 | |
XM_006240401.2 | XP_006240463.1 | 4 - 5 | 651 | 64 | ||
XM_006240402.2 | XP_006240464.1 | 4 - 5 | 848 | 64 | ||
XM_006240403.2 | XP_006240465.1 | 4 - 5 | 981 | 64 | ||
XM_008764893.1 | XP_008763115.1 | 5 - 6 | 732 | 64 | ||
XM_008764894.1 | XP_008763116.1 | 5 - 6 | 946 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
