Assay Details
Gene Symbol: Asb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011984.1 | NP_001011984.1 | 6 - 7 | 1057 | 55 | |
XM_006240464.2 | XP_006240526.1 | 4 - 5 | 1702 | 55 | ||
GenBank mRNA | BC085882.1 | - | 6 - 7 | 1057 | 7191983 | 55 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
