Assay Details
Gene Symbol: Psen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019163.3 | NP_062036.2 | 3 - 4 | 487 | 82 | |
XM_006240321.2 | XP_006240383.1 | 5 - 6 | 757 | 82 | ||
XM_006240322.2 | XP_006240384.1 | 5 - 6 | 1010 | 82 | ||
GenBank mRNA | BC070887.1 | - | 5 - 6 | 683 | 82 | |
D82363.1 | - | 3 - 4 | 487 | 82 | ||
D82578.1 | - | 3 - 4 | 487 | 82 | ||
FQ231303.1 | - | 5 - 6 | 705 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
