Assay Details
Gene Symbol: Psen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019163.3 | NP_062036.2 | 8 - 9 | 1134 | 90 | |
XM_006240321.2 | XP_006240383.1 | 10 - 11 | 1404 | 90 | ||
XM_006240322.2 | XP_006240384.1 | 10 - 11 | 1657 | 90 | ||
GenBank mRNA | BC070887.1 | - | 10 - 11 | 1330 | 90 | |
D82578.1 | - | 8 - 9 | 1134 | 90 | ||
FQ231303.1 | - | 10 - 11 | 1352 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
