Assay Details
Gene Symbol: Srsf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195505.1 | NP_001182434.1 | 251 | 70 | ||
XM_008764878.1 | XP_008763100.1 | 1 - 2 | 302 | 70 | ||
GenBank mRNA | FM049719.1 | - | 1 - 2 | 214 | 70 | |
L13635.1 | - | 222 | 70 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the L13635.1, NM_001195505.1, XM_008764878.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
