Assay Details
Gene Symbol: Dcaf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108716.1 | NP_001102186.1 | 1 - 2 | 124 | 73 | |
XM_006240347.2 | XP_006240409.1 | 1 - 2 | 124 | 73 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001108716.1, XM_006240347.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
