Assay Details
Gene Symbol: Ptgr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015009.1 | NP_001015009.1 | 5 - 6 | 684 | 57 | |
XM_006240326.1 | XP_006240388.1 | 5 - 6 | 698 | 57 | ||
XM_006240327.1 | XP_006240389.1 | 5 - 6 | 634 | 57 | ||
XM_006240328.2 | XP_006240390.1 | 5 - 6 | 596 | 57 | ||
XM_006240329.2 | XP_006240391.1 | 3 - 4 | 1221 | 57 | ||
GenBank mRNA | BC091173.1 | - | 5 - 6 | 684 | 7189599 | 57 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
