Assay Details
Gene Symbol: Alkbh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108718.1 | NP_001102188.1 | 3 - 4 | 466 | 82 | |
XM_006240394.2 | XP_006240456.1 | 3 - 4 | 472 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
