Assay Details
Gene Symbol: Alkbh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108718.1 | NP_001102188.1 | 4 - 5 | 559 | 78 | |
GenBank mRNA | FQ226790.1 | - | 1 - 2 | 126 | 78 | |
FQ232964.1 | - | 3 - 4 | 235 | 78 | ||
FQ234582.1 | - | 1 - 2 | 126 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
