Assay Details
Gene Symbol: Alkbh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108718.1 | NP_001102188.1 | 1 - 2 | 205 | 77 | |
XM_006240394.2 | XP_006240456.1 | 1 - 2 | 211 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
