Assay Details
Gene Symbol: Itpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191985.1 | NP_001178914.1 | 3 - 4 | 243 | 123 | |
XM_008764841.1 | XP_008763063.1 | 2 - 3 | 1111 | 123 | ||
XM_008764842.1 | XP_008763064.1 | 2 - 3 | 1582 | 123 | ||
XM_008764843.1 | XP_008763065.1 | 2 - 3 | 4089 | 123 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008764843.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
