Assay Details
Gene Symbol: Itpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191985.1 | NP_001178914.1 | 9 - 10 | 897 | 68 | |
XM_008764841.1 | XP_008763063.1 | 9 - 10 | 1891 | 68 | ||
XM_008764842.1 | XP_008763064.1 | 9 - 10 | 2362 | 68 | ||
XM_008764843.1 | XP_008763065.1 | 9 - 10 | 4869 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
