Assay Details
Gene Symbol: Gnai3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013106.1 | NP_037238.1 | 5 - 6 | 666 | 64 | |
GenBank mRNA | DQ120473.1 | - | 5 - 6 | 559 | 64 | |
DQ120474.1 | - | 5 - 6 | 559 | 64 | ||
FQ222046.1 | - | 5 - 6 | 729 | 64 | ||
J03219.1 | - | 5 - 6 | 675 | 64 | ||
M20713.1 | - | 5 - 6 | 666 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
