Assay Details
Gene Symbol: Ube2d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233333.1 | XP_006233395.1 | 8 - 9 | 717 | 86 | |
XM_006233334.1 | XP_006233396.1 | 8 - 9 | 864 | 86 | ||
XM_006233335.1 | XP_006233397.1 | 8 - 9 | 671 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
