Assay Details
Gene Symbol: Extl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100704.1 | NP_001094174.1 | 2 - 3 | 187 | 94 | |
XM_006233215.2 | XP_006233277.1 | 2 - 3 | 732 | 94 | ||
XM_006233216.2 | XP_006233278.1 | 2 - 3 | 473 | 94 | ||
GenBank mRNA | BC162014.1 | - | 2 - 3 | 187 | 94 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC162014.1, NM_001100704.1, XM_006233215.2, XM_006233216.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
