Assay Details
Gene Symbol: Bdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106473.1 | NP_001099943.1 | 9 - 10 | 907 | 79 | |
XM_006233322.2 | XP_006233384.1 | 9 - 10 | 816 | 79 | ||
GenBank mRNA | BC095848.1 | - | 8 - 9 | 1301 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
