Assay Details
Gene Symbol: Ddost | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012104.1 | NP_001012104.1 | 5 - 6 | 558 | 74 | |
GenBank mRNA | BC082075.1 | - | 5 - 6 | 558 | 74 | |
FQ228905.1 | - | 5 - 6 | 590 | 74 | ||
FQ229122.1 | - | 5 - 6 | 570 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
