Assay Details
Gene Symbol: Ccnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013094.1 | NP_001013112.1 | 2 - 3 | 356 | 73 | |
XM_006239544.2 | XP_006239606.1 | 2 - 3 | 878 | 73 | ||
XM_006239545.2 | XP_006239607.1 | 2 - 3 | 877 | 73 | ||
GenBank mRNA | BC088316.1 | - | 2 - 3 | 356 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
