Assay Details
Gene Symbol: Masp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172043.1 | NP_742040.1 | 8 - 9 | 1120 | 52 | |
GenBank mRNA | AJ277747.1 | - | 7 - 8 | 1062 | 52 | |
BC128700.1 | - | 8 - 9 | 1121 | 52 | ||
Y18564.1 | - | 7 - 8 | 899 | 52 | ||
Y18565.1 | - | 1 - 2 | 45 | 52 | ||
Y18566.1 | - | 5 - 6 | 652 | 52 | ||
Y18571.1 | - | 2 - 3 | 127 | 52 | ||
Y18572.1 | - | 7 - 8 | 957 | 52 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
