Assay Details
Gene Symbol: Pink1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106694.1 | NP_001100164.1 | 2 - 3 | 707 | 84 | |
GenBank mRNA | BC169047.1 | - | 2 - 3 | 701 | 7190543 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
