Assay Details
Gene Symbol: Dlgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276304.1 | NP_001263233.1 | 7 - 8 | 2131 | 56 | |
XM_008763989.1 | XP_008762211.1 | 8 - 9 | 3433 | 56 | ||
GenBank mRNA | AY530298.1 | - | 6 - 7 | 1994 | 56 | |
AY530299.1 | - | 6 - 7 | 1994 | 56 | ||
U67139.1 | - | 7 - 8 | 2098 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
