Assay Details
Gene Symbol: Gpr157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012107.1 | NP_001012107.1 | 1 - 2 | 616 | 68 | |
GenBank mRNA | BC090013.1 | - | 1 - 2 | 616 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
