Assay Details
Gene Symbol: Tnfrsf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025773.1 | NP_001020944.1 | 6 - 7 | 981 | 67 | |
XM_006239471.2 | XP_006239533.1 | 7 - 8 | 1401 | 67 | ||
XM_006239472.2 | XP_006239534.1 | 6 - 7 | 1834 | 67 | ||
XM_008764282.1 | XP_008762504.1 | 6 - 7 | 1171 | 67 | ||
XM_008764283.1 | XP_008762505.1 | 7 - 8 | 1422 | 67 | ||
GenBank mRNA | AB164643.1 | - | 5 - 6 | 557 | 67 | |
BC097483.1 | - | 6 - 7 | 981 | 7459228 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
