Assay Details
Gene Symbol: Psmb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017284.1 | NP_058980.1 | 2 - 3 | 264 | 115 | |
GenBank mRNA | BC058487.1 | - | 2 - 3 | 284 | 115 | |
D21799.1 | - | 2 - 3 | 264 | 115 | ||
FQ217737.1 | - | 2 - 3 | 294 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
