Assay Details
Gene Symbol: Mrps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007653.1 | NP_001007654.1 | 3 - 4 | 330 | 97 | |
GenBank mRNA | BC083856.1 | - | 3 - 4 | 330 | 7125450 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
