Assay Details
Gene Symbol: Rcan3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239218.2 | XP_006239280.1 | 1 - 2 | 240 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006239218.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
