Assay Details
Gene Symbol: Serinc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031656.1 | NP_001026826.1 | 3 - 4 | 436 | 133 | |
GenBank mRNA | BC099093.1 | - | 3 - 4 | 436 | 7457983 | 133 |
DQ103709.1 | - | 3 - 4 | 422 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
