Assay Details
Gene Symbol: Serinc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031656.1 | NP_001026826.1 | 5 - 6 | 654 | 73 | |
GenBank mRNA | BC099093.1 | - | 5 - 6 | 654 | 7457983 | 73 |
DQ103709.1 | - | 5 - 6 | 640 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
