Assay Details
Gene Symbol: Fgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024145.2 | NP_077059.2 | 10 - 11 | 1328 | 94 | |
XM_006239078.2 | XP_006239140.1 | 9 - 10 | 1228 | 94 | ||
XM_006239079.2 | XP_006239141.1 | 10 - 11 | 1260 | 94 | ||
XM_006239080.2 | XP_006239142.1 | 10 - 11 | 1260 | 94 | ||
XM_006239081.2 | XP_006239143.1 | 10 - 11 | 1341 | 94 | ||
XM_006239082.2 | XP_006239144.1 | 9 - 10 | 1272 | 94 | ||
GenBank mRNA | BC062025.1 | - | 10 - 11 | 1328 | 5623501 | 94 |
X57018.1 | - | 10 - 11 | 1269 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
