Assay Details
Gene Symbol: Rcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128189.2 | NP_001121661.1 | 9 - 10 | 1096 | 59 | |
XM_006239073.2 | XP_006239135.1 | 9 - 10 | 1066 | 59 | ||
XM_006239074.1 | XP_006239136.1 | 9 - 10 | 1390 | 59 | ||
XM_006239075.2 | XP_006239137.1 | 9 - 10 | 1580 | 59 | ||
XM_006239077.2 | XP_006239139.1 | 8 - 9 | 1026 | 59 | ||
XM_008764172.1 | XP_008762394.1 | 9 - 10 | 2734 | 59 | ||
XM_008764173.1 | XP_008762395.1 | 10 - 11 | 1120 | 59 | ||
GenBank mRNA | BC160834.1 | - | 9 - 10 | 1096 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
