Assay Details
Gene Symbol: Rcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128189.2 | NP_001121661.1 | 6 - 7 | 701 | 69 | |
XM_006239073.2 | XP_006239135.1 | 6 - 7 | 671 | 69 | ||
XM_006239074.1 | XP_006239136.1 | 6 - 7 | 995 | 69 | ||
XM_006239075.2 | XP_006239137.1 | 6 - 7 | 1185 | 69 | ||
XM_006239077.2 | XP_006239139.1 | 5 - 6 | 631 | 69 | ||
XM_008764172.1 | XP_008762394.1 | 6 - 7 | 2339 | 69 | ||
XM_008764173.1 | XP_008762395.1 | 7 - 8 | 725 | 69 | ||
GenBank mRNA | BC160834.1 | - | 6 - 7 | 701 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
