Assay Details
Gene Symbol: Rcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128189.2 | NP_001121661.1 | 2 - 3 | 158 | 75 | |
GenBank mRNA | BC160834.1 | - | 2 - 3 | 158 | 7456254 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC160834.1, NM_001128189.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
