Assay Details
Gene Symbol: Ppp1r8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107911.1 | NP_001101381.1 | 4 - 5 | 506 | 66 | |
GenBank mRNA | BC162025.1 | - | 4 - 5 | 505 | 9099457 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
