Assay Details
Gene Symbol: Mad2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012106.1 | NP_001012106.1 | 3 - 4 | 304 | 67 | |
XM_006239390.2 | XP_006239452.1 | 5 - 6 | 473 | 67 | ||
XM_006239391.2 | XP_006239453.1 | 3 - 4 | 989 | 67 | ||
XM_008764256.1 | XP_008762478.1 | 4 - 5 | 710 | 67 | ||
GenBank mRNA | BC087687.1 | - | 3 - 4 | 304 | 7307981 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
