Assay Details
Gene Symbol: Mad2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012106.1 | NP_001012106.1 | 6 - 7 | 646 | 71 | |
XM_006239390.2 | XP_006239452.1 | 8 - 9 | 815 | 71 | ||
XM_006239391.2 | XP_006239453.1 | 5 - 6 | 1185 | 71 | ||
XM_008764256.1 | XP_008762478.1 | 7 - 8 | 1052 | 71 | ||
GenBank mRNA | BC087687.1 | - | 6 - 7 | 646 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
