Assay Details
Gene Symbol: Mtfr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013935.1 | NP_001013957.1 | 2 - 3 | 144 | 74 | |
XM_006239124.2 | XP_006239186.1 | 2 - 3 | 440 | 74 | ||
XM_006239125.2 | XP_006239187.1 | 2 - 3 | 218 | 74 | ||
GenBank mRNA | BC083692.1 | - | 2 - 3 | 144 | 7189448 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC083692.1, NM_001013935.1, XM_006239124.2, XM_006239125.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
