Assay Details
Gene Symbol: Mtfr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239125.2 | XP_006239187.1 | 1 - 2 | 95 | 57 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006239125.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
