Assay Details
Gene Symbol: Gnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030987.2 | NP_112249.2 | 11 - 12 | 1177 | 63 | |
XM_006239517.2 | XP_006239579.1 | 11 - 12 | 1329 | 63 | ||
XM_008764335.1 | XP_008762557.1 | 10 - 11 | 1212 | 63 | ||
GenBank mRNA | AF022083.1 | - | 1 - 2 | 117 | 63 | |
BC078809.1 | - | 11 - 12 | 1177 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
G protein-coupled receptor signaling pathway
phospholipase C-activating G protein-coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
metabolic process
cell population proliferation
cardiac muscle cell apoptotic process
sensory perception of taste
retina development in camera-type eye
cellular response to hypoxia
G protein-coupled receptor binding
GTPase activity
protein binding
spectrin binding
type 1 angiotensin receptor binding
protein-containing complex binding
GTPase binding
G protein-coupled receptor signaling pathway
phospholipase C-activating G protein-coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
metabolic process
cell population proliferation
cardiac muscle cell apoptotic process
sensory perception of taste
retina development in camera-type eye
cellular response to hypoxia
G protein-coupled receptor binding
GTPase activity
protein binding
spectrin binding
type 1 angiotensin receptor binding
protein-containing complex binding
GTPase binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P384857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946253 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159316 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783271 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826533 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345169 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134507 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361209 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532650 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167768 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704367 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144474 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861887 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71290 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163457 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91213 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173930 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P384857 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P384857 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P384857 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P468372 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P468365 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P927153 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P13986 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P802937 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700515072 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700772122 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 64433 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 62088 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 24548 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 90700 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700515072 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700772122 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10874650 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10874653 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10874648 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10874643 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10874649 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788438 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10874644 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10874645 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10874651 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10874646 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10874647 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10874642 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10874652 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10788437 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10874641 | - | - |
Rat Genome 230 2.0 Array | 1367731_PM_at | - | - |
Rat Genome 230 2.0 Array | 1367731_at | - | - |
Rat Genome 230 2.0 Array | 1375705_PM_at | - | - |
Rat Genome 230 2.0 Array | 1367732_at | - | - |
Rat Genome 230 2.0 Array | 1375705_at | - | - |
Rat Genome 230 2.0 Array | 1367732_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1367731_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1367732_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1367732_at | - | - |
Affymetrix HT Rat Focus Array | 1367731_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500015660.rn.1 | TC0500002063.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500015658.rn.1 | TC0500002063.rn.1 | RTA TC |
Rat Expression Set 230 | 1367731_at | - | - |
Rat Expression Set 230 | 1367731_PM_at | - | - |
Rat Expression Set 230 | 1367732_at | - | - |
Rat Expression Set 230 | 1367732_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10788437 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10874641 | - | - |
Rat Genome U34 Set | AF022083_s_at | - | - |
Rat Genome U34 Set | U88324_at | - | - |
Rat Genome U34 Set | U88324_g_at | - | - |
Rat Genome U34 Set | rc_AI103622_at | - | - |
Rat Genome U34 Set | rc_AI230404_s_at | - | - |
Rat Genome U34 Set | rc_AI227660_s_at | - | - |
Rat Genome U34 Set | U88324_at | - | - |
Rat Genome U34 Set | AF022083_s_at | - | - |
Rat Genome U34 Set | rc_AI227660_s_at | - | - |
Rat Genome U34 Set | U88324_g_at | - | - |
Rat Genome U34 Set | rc_AI230404_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22158285 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1375069 | - | - |
RatRef-12 Expression BeadChip | ILMN_49575 | - | - |
