Assay Details
Gene Symbol: Ptp4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC060549.1 | - | 1 - 2 | 581 | 63 | |
FQ230760.1 | - | 1 - 2 | 91 | 63 | ||
FQ231532.1 | - | 1 - 2 | 215 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
