Assay Details
Gene Symbol: Pabpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238798.1 | XP_006238860.1 | 10 - 11 | 2360 | 80 | |
XM_006238799.1 | XP_006238861.1 | 10 - 11 | 2360 | 80 | ||
GenBank mRNA | BC088337.1 | - | 5 - 6 | 719 | 80 |
Gene Symbol: RGD1562451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769025.1 | XP_008767247.1 | 1 | 1794 | 80 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
