Assay Details
Gene Symbol: Pabpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100538.1 | NP_001094008.1 | 2 - 3 | 753 | 85 | |
XM_006238798.1 | XP_006238860.1 | 2 - 3 | 1293 | 85 | ||
XM_006238799.1 | XP_006238861.1 | 2 - 3 | 1293 | 85 | ||
XM_006238800.1 | XP_006238862.1 | 2 - 3 | 1293 | 85 | ||
XM_006238801.1 | XP_006238863.1 | 2 - 3 | 1293 | 85 | ||
GenBank mRNA | BC166452.1 | - | 2 - 3 | 758 | 85 |
Gene Symbol: RGD1562451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769025.1 | XP_008767247.1 | 1 | 727 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
